Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Valproic Acid, USP, 98-102%, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Valproic Acid, USP is used to treat certain types of seizures. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
Ignition Residue | 0.001 |
Packaging | Amber Glass Bottle |
Quantity | 100 g |
Percent Purity | 98 to 102% |
Grade | USP |