Learn More
Medchemexpress LLC Valrocemide | 92262-58-3 | MFCD00870963 | 99.4% | 200.28 g/mol | C10H20N2O2 | 25 MG

Supplier: Medchemexpress LLC HY10037925MG
Valrocemide is a small-molecule anticonvulsant research compound described as a promising antiepileptic candidate with broad-spectrum anticonvulsant activity. It is supplied as a white to off-white solid for laboratory research use only and is characterized by a reported purity of 99.42%.
- Broad-spectrum anticonvulsant activity reported in preclinical studies.
- High reported purity (99.42%).
- White to off-white solid suitable for analytical and in vivo research.
- Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
- Solvent stability for prepared solutions: -80°C (2 years) or -20°C (1 year).
- Available in small research quantities, useful for assay development and pilot studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.