Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ValRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159311AP

Catalog No. 50-173-0581


Add to cart

Description

Description

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. The protein encoded by this gene belongs to class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex.
Specifications

Specifications

ValRS
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26640, Q04462, Q9Z1Q9
G7A, Protein G7a, Valine tRNA ligase, ValRS, Valyl-tRNA synthetase, VARS, VARS1, VARS2
Rabbit
IgG
150 μL
-20°C
Primary
22321, 25009, 7407
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
VARS
Liquid
VARS
ValRS Fusion Protein Ag8778
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit