Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Lab Refrigerator, 5.5 cu. ft., 115V PROMO

Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators, featuring adjustable temperature control and manual defrost.

Manufacturer:  Thermo Scientific™ 05LREETSARP

Catalog No. 05-LRE-ETSARP

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

  • Adjustable shelves or baskets included
  • Environmentally friendly, CFC-free insulation and coolant
  • Factory tested prior to shipment
  • NRTL tested
  • Easy-to-clean, durable finish
  • Specifications

    Specifications

    1°C to 12°C
    5.5 cu. ft.
    115 V 60 Hz
    60 Hz
    NEMA 5-15 P
    17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm)
    54.5 kg
    ABS Plastic
    13 Months Parts and Labor
    Enameled Steel
    10 Years
    Interior - Yes
    2 Adjustable Feet In Front, 2 Fixed Pads In Rear
    Manual
    Mechanical Dial Thermostat/ Inside, Upper Right Side
    Bottom Mounted
    1 (1 compartment)
    3 Adjustable Shelves, Storage On Bottom Of Unit
    cULus
    155.7 L
    115 V
    1.0 A
    23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1 cm)
    100 lb.
    R134a, 100% CFC Free
    Door Lock
    White
    Indoor General Storage Of Non-Critical Samples And Reagents
    2 in. On Sides, 2 in. At Back, 2 in. On Top
    Adjustable
    05LREETSA
    General Purpose Refrigerator
    Hinged, Open Left to Right, Reversible
    Polyurethane Foam
    160 w
    SDS
    Documents
    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit