Value Lab Refrigerators

Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators.

Manufacturer: Thermo Scientific 20LREETSA

 View more versions of this product

Catalog No. 20LREETSA

  Check Availability
Add to Cart

Alert

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.


    Specifications

    Access Security No door lock
    Dimensions (L x W x H) Exterior 31.25 x 33 x 67.25 in. (79.4 x 83.8 x 170.8cm)
    Dimensions (D x W x H) Interior 23.5 x 27 x 56” (59.7 x 68.6 x 142.2 cm)
    Temperature Range (Metric) 1°C to 12°C,1° to 12°C
    Min. Clearance 3 in. On Sides, 3 in. At Back, 3 in. On Top
    Doors 1 (1 compartment)
    For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
    Typical Lifetime 10 Years
    Lighting Interior - Yes
    Temperature (Setpoint) Adjustable Set Point - Yes
    Color White
    Control Mechanical Dial Thermostat/ Lower Right Back Wall
    Warranty 13 Months Parts and Labor
    Current 4.0 Amp                                              
    Compressor Bottom Mounted
    Exterior Finish Enameled Steel
    Interior Finish Enameled steel,HIPS polystyrene liner
    Model 20LREETSA
    Type General Purpose Refrigerator
    Voltage 115V,120V
    Certifications/Compliance cCSAus
    Electrical Requirements 115 V/60 Hz
    Amperage 4.0A
    Capacity 20.3 Cubic Feet/574.8 Liters,20 cu. ft. / 566.34L
    Capacity (English) 20 cu. ft.,20 cu. ft.
    Capacity (Metric) 566.34L,566.34L
    Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear
    Defrost Manual,Manual Defrost
    Door Count 1 (1 Compartment),1(1 compartment)
    Door Style Hinged, Open Left to Right, Non-Reversible,Hinged, Opens Left to Right, Non-Reversible
    Hertz 60Hz
    Insulation Polyurethane Foam
    Plug Type NEMA 5-15 P,Yes/Three prong NEMA 5-15P, 3.048 Meters (10 Feet)
    Shipping Weight (English) 283 lb.
    Shelves 4 fixed Interior shelves, 1 Wire Basket, 6 door shelves,4 Fixed Shelves, 6 Door Shelves
    Refrigerant R134A, CFC Free,R134A,CFCFree
    Shipping Weight (Metric) 128kg
    Width (English) Exterior 33 in.
    Chamber Material Painted steel, White
    Depth (Metric) Interior 59.7cm
    Height (English) Exterior 67.25 in.
    Height (English) Interior 56 in.
    Height (Metric) Exterior 170.8cm
    Height (Metric) Interior 142.2cm
    Length (English) Exterior 31.25 in.
    Width (English) Interior 27 in.
    Width (Metric) Exterior 83.8cm
    Depth (English) Interior 23.5 in.

    Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators, featuring adjustable temperature control and manual defrost.

    • Adjustable shelves or baskets included
    • Environmentally friendly, CFC-free insulation and coolant
    • Factory tested prior to shipment
    • NRTL tested
    • Easy-to-clean, durable finish 

    Ordering Information:

    Not available outside North America.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit