Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Lab Refrigerators

Store samples safely at temperatures from 1° to 12°C with these value-oriented laboratory refrigerators.

Manufacturer:  Thermo Scientific™ 05LREETSV

 View more versions of this product

Catalog No. 05LREETSV

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Alert:

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators, featuring adjustable temperature control and manual defrost.

  • Adjustable shelves or baskets included
  • Environmentally friendly, CFC-free insulation and coolant
  • Factory tested prior to shipment
  • NRTL tested
  • Easy-to-clean, durable finish 

Ordering Information:

Not available outside North America.

Specifications

Specifications

1°C to 8°C
155,7 L
230 V
0.5 A
23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1 cm)
100 lb.
100 lb.
R134a, 100% CFC Free
Door Lock
White
Indoor General Storage Of Non-Critical Samples And Reagents
2 in. On Sides, 2 in. At Back, 2 in. On Top
Adjustable
05LREETSV
General Purpose Refrigerator
Hinged, Opens Left to Right, Reversible
Polyurethane Foam
1 (1 compartment)
3 Adjustable Shelves, Storage On Bottom Of Unit
5.5 cu. ft.
230 V 50 Hz
50 Hz
NEMA 6-15 P
17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm)
54.5 kg
45.3 kg
ABS Plastic
13 Months Parts and Labor
Enameled Steel
10 Years
Interior - Yes
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Manual
Mechanical Dial Thermostat/ Inside, Upper Right Side
Bottom Mounted
ABS Plastic (Interior)
120 w
Undercounter
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit