Thermo Scientific™ Value Refrigerator/Freezer Available on GSA/VA Contract for Federal Government customers only.

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Manufacturer: Thermo Scientific™ 25LCEETSA

 View more versions of this product

Catalog No. 25LCEETSA

  Check Availability
Add to Cart

Alert

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.


    Specifications

    Warranty Warranty varies by country, please contact us for details
    Capacity 24.9 Cubic Feet/705.1 Liters
    Capacity (English) 24.9 cu. ft.
    Capacity (Metric) 705.1 L
    Dimensions (L x W x H) Exterior 37.5 x 32.75 x 69.75 in. (95.2 x 83.19 x 177.17cm)
    Dimensions (D x W x H) Interior
    Frz: 29.2 x 30.2 x 20.6 in. (74.1 x 76.8 x 52.3 cm)
    Ref: 29.2 x 30.2 x 40.6 in. (74.1 x 76.8 x 101.3 cm)
    Shipping Weight (English) 275 lbs
    Shipping Weight (Metric) 124.7 kg
    Temperature Range (Metric) Ref: 1°C to 12°C Frz: -14°C to -25°C
    Temperature Range (Freezer) -14°C to -25°C
    Type Combination Refrigerator/Freezer
    Min. Clearance .125 in. On Sides, 2 in. At Back, 1 in. On Top
    Temperature (Setpoint) Adjustable
    Display Temperature - None
    For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
    Typical Lifetime 10 Years
    Lighting Interior - Yes
    Current 11.6 Amps
    Model 25LCEETSA
    Control Digital/Inside Top Front Of Fridge
    Door Count 2(2 compartments)
    Exterior Finish Epoxy Enameled Steel
    Interior Finish ABS Plastic
    Color White
    Certifications/Compliance cULus
    Electrical Requirements 115 V/60 Hz
    Defrost Automatic Defrost
    Plug Type Yes/Three prong NEMA 5-15P/ 3.048 Meters (10 Feet)
    Shelves Ref: 3 Shelves, 4 Door Half Shelves and 2 Storage Bins; Frzr: 1 Shelf And 3 Wire Baskets
    Amperage 11.6A
    Hertz 60Hz
    Access Security Door lock
    Temperature Range (Refrigerator) 1° to 12°C
    Compressor Bottom Mounted
    Door Style Hinged, Opens Left to Right, Reversible
    Voltage 115V
    Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear
    Insulation Polyurethane Foam
    Refrigerant R134A, CFC Free

    Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

    Features:

    • Factory tested prior to shipment
    • NRTL listed
    • Environmentally-friendly, non CFC refrigerants
    • Adjustable temperature control
    • Easy to clean, durable finish
    • Manual and auto defrost combination units

    Ordering Information:

    • Not available outside North America

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit