Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Refrigerator/Freezer

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Manufacturer:  Thermo Scientific™ 05LCEETSA/DEL

 View more versions of this product

Catalog No. 05LCEETSA

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Alert:

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.


Description

Description

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Features:
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, non CFC refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto defrost combination units

Ordering Information:

  • Not available outside North America
Specifications

Specifications

1°C to 12°C (Refrigerator), -12°C to -20°C (Freezer)
5.6 cu. ft.
115 V 60 Hz
60 Hz
NEMA 5-15 P
Ref: 17 x 20.5 x 19.5 in. (43.2 x 52.1 x 49.5 cm)
Frz: 11.5 x 19 x 5 in. (29.2 x 48.3 x 12.7 cm)
46 kg
55 kg
Polyurethane Foam
Hinged, Opens Left to Right, Reversible
2 Adjustable Shelves, 3 Door Shelves, 1 Drawer
Warranty varies by country, please contact us for details
Enameled Steel
Indoor General Storage Of Non-Critical Samples And Reagents
2 in. On Sides, 2 in. At Back, 2 in. On Top
Adjustable
Bottom Mounted
-12°C to -20° C
1°C to 12°C
Mechanical Dial Thermostat/Inside Upper Right Side
cULus
158.6 L
115 V
1.3 A
23.5 x 23.63 x 33.5 in. (61 x 64.8 x 85.1 cm)
100 lb.
120 lb.
Temperature Display - None
R134a, 100% CFC Free
ABS plastic
Door Lock
White
160 w
10 Years
Interior - Yes
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
05LCEETSA
Manual
Combination General Purpose Refrigerator/Freezer
1 (2 compartments)
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit