Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Refrigerator/Freezer

Chill, freeze and retrieve samples safely, simply and economically with this value-oriented refrigerator/freezer.

Manufacturer:  Thermo Scientific™ 05LCEETSV

 View more versions of this product

Catalog No. 05LCEETSV

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Alert:

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Features:
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, non CFC refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto defrost combination units

Ordering Information:

  • Not available outside North America
Specifications

Specifications

1°C to 12°C (Refrigerator), -12°C to -20°C (Freezer)
158,6 L
230 V
0.5 A
23.5 x 23.63 x 33.5 in. (61 x 64.8 x 85.1 cm)
100 lb.
120 lb.
Temperature Display - None
ABS plastic
Warranty varies by country, please contact us for details
Enameled Steel
10 Years
Interior - Yes
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
-12°C to -20°C
1°C to 12°C
Mechanical Dial Thermostat/Inside Upper Right Side
Bottom Mounted
1 (2 compartments)
120 w
5.6 cu. ft.
230 V 50 Hz
50 Hz
NEMA 6-15 P
Ref: 17 x 20.5 x 19.5 in. (43.2 x 52.1 x 49.5 cm)
Frz: 11.5 x 19 x 5 in. (29.2 x 48.3 x 12.7 cm)
46 kg
55 kg
R134a, 100% CFC Free
Door Lock
White
Indoor General Storage Of Non-Critical Samples And Reagents
2 in. On Sides, 2 in. At Back, 2 in. On Top
Adjustable
05LCEETSV
Manual
Combination General Purpose Refrigerator/Freezer
Hinged, Opens Left to Right, Reversible
Polyurethane Foam
6
2 Adjustable Shelves, 3 Door Shelves, 1 Drawer
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit