Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Refrigerator/Freezer Available on GSA/VA Contract for Federal Government customers only.

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Manufacturer:  Thermo Scientific™ 18LCEETSA

 View more versions of this product

Catalog No. 18LCEETSA


Add to cart

Alert:

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.


Description

Description

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Features:
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, non CFC refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto defrost combination units

Ordering Information:

  • Not available outside North America

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0°C to 8°C (Refrigerator), -6°C to -20°C (Freezer)
18.2 cu. ft.
115 V 60 Hz
60 Hz
NEMA 5-15 P
Ref: 24.75 x 26.0 x 39.75 in. (62.86 x 66.04 x 100.9 cm)
Frz: 21.0 x 26.0 x 15.5 in. (53.34 x 66.04 x 39.4 cm)
108.8 kg
93 kg
Polyurethane Foam
Hinged, Open Left to Right, Reversible
(2) Adjustable (1) Fixed Shelves, (3) Door Shelves & (2) Drawers In Refrigerator, (2) Door Shelves & (1) Shelf In Freezer
Warranty varies by country, please contact us for details
Enameled Steel
Indoor General Storage Of Non-Critical Samples And Reagents
.375 in. On Sides, 1 in. At Back, .375 in. On Top
Adjustable
2 Fixed Rollers In Front, 2 Rollers In Rear
18LCEETSA
Automatic
Combination General Purpose Refrigerator/Freezer
2 (2 compartments)
cCSAus
515.4 L
115 V
6 A
30.13 x 30 x 66.63 in. (76.5 x 76.2 x 169.24 cm)
240 lb.
205 lb.
Temperature Display - None
R134a, 100% CFC Free
ABS Plastic
None
White
690 w
10 Years
Interior - Yes
Epoxy-enameled Steel
Bottom Mounted
-12°C to -20° C
1°C to 12°C
Mechanical Dial Thermostat/Inside, Top Of Refrigerator
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit