VALUE TEK Disposable 5 Snap Labcoats

Manufacturer: VALUE TEK VTLBCT3X

 View more versions of this product

Catalog No. 19-150-0840

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Disposable Yes
    Cuff Style Elastic
    Material Spunbond Polypropylene
    No. of Pockets 3.0
    Size 3X-Large
    For Use With (Application) Used in electronic medical device and pharmaceuticals industries
    Certifications/Compliance ISO 7
    Closure Type Snap
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit