VALUE TEK Nylon Glove Liner

Manufacturer: VALUE TEK VTGNLR1/1

 View more versions of this product

Catalog No. 19-150-0834

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Launderable
    Finger Style Full
    Hand Ambidextrous
    For Use With (Application) use in controlled environment, limits Skin irritation
    Laboratory/Cleanroom Use Yes
    Material Nylon
    Size Universal
    Weight Light weight
    Glove Cuff Color White
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit