Learn More
ValueMax™ Extra-Safe™ Knee-Length Lab Coat Medical Blue
Manufacturer:  ValueMax™ NP3660MB2X
 View more versions of this product
Catalog No. 19-636-014
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Knee-Length
  • Latex-free
  • Fluid-resistant, breathable 3-layer SMS fabric
  • Snap front
  • No pockets
  • Knit collar
  • Autoclavable
  • Washable up to
  • 5 times Blue
Specifications
Specifications
Safety
Snaps
Knit
2X-Large
3-ply SMS
OSHA
Blue
SMS
Yes