Learn More
Fisherbrand™ ValuGard™ Crown Face Shield with Headgear
Contour bar reduces eyeglass fogging
Catalog No. 01-361-92
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Blue nonwoven synthetic fiber outer layer surrounds fiberglass-free polyfiltration inner layer.
  • Bacteria filtration exceeds 92% at 1.0μm
  • Three-ply construction with two sets of Tyvek™ ties
  • Surgical style
  • ISO Class 5 compatible
Specifications
Specifications
3-ply
Surgical
Non-woven Synthetic Fiber outer layer surrounds Fiberglass-free polyfiltration inner layer
Synthetic
Standard
92% (BFE)
Blue
Two sets of Tyvek™ ties, polyfiltration medium
1 (BFE)μ