Learn More
ValueMax™ Easy-Breathe™ Lab Coats - Ceil Blue
Click to view available options
Size:
Small
Medium
4X-Large
5X-Large
Description
- Silky soft “SMS” fabric which meets AAMI level 2 fluid resistant standards
- Anti-static treated and clinging-free
- Knee length lab coats with snap front and three pockets
- Knitted cuffs and collar
- Economical
- Color: Ceil Blue
Specifications
Size | Medium |
Material | SMS |
Color | Ceil Blue |
Cuff Style | Knit |
Collar Type | Knit |