Learn More
Valuemax™ Easy-Breathe™ Lab Coats - Medical Blue
Combination of fluid resistance and breathability
Manufacturer:  Valuemax™ 3560MB3XL
 View more versions of this product
Catalog No. 23-900-761
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Silky soft “SMS” fabrics which meets AAMI level 2 fluid resistant standards
  • Anti-static treated and clinging-free
  • Upto knee length, snap front and has three pockets
  • Knitted cuffs and collar
  • Economical
  • Color:Medical Blue
Specifications
Specifications
Knitted
Knit
3X-Large
Medical Blue
Silky soft SMS fabric