Learn More
Valuemax™ Easy-Breathe™ Lab Coats - White
Great combination of fluid resistance and breathability
Manufacturer:  Valuemax™ 3560WHM
 View more versions of this product
Catalog No. 23-900-789
Add to cart
Description
Description
  • Silky soft “SMS” fabric which meets AAMI level 2 fluid resistant standards
  • Anti-static treated and clinging-free
  • Knee length lab coats with snap front and three pockets
  • Knitted cuffs and collar
  • Economical
  • Color: White
Specifications
Specifications
Knitted
Knit
Medium
White
Silky soft SMS fabric