Learn More
Valuemax™ Easy-Breathe™ Lab Jackets - Medical Blue
Affordable apparels combined with fluid resistance and breathability
Manufacturer:  Valuemax™ 3530MB2XL
 View more versions of this product
Catalog No. 23-900-712
Add to cart
Description
Description
  • Silky soft “SMS” fabrics
  • Anti-static treated and no clinging
  • Snap front, three pockets
  • Knit cuff and collar
  • Hip length and bright colors for any operations
  • Meet AAMI level 2 fluid resistant standard
Specifications
Specifications
Knit
Medical Blue
Knit
2X-Large