ValuMax™ Extra-Safe™ Lab Coats — Medical Blue
Static-free — no clinging
Manufacturer: Valumax International, Inc 3660MBXL
 View more versions of this product
Catalog No. 23-900-521D
  Check Availability
Add to Cart
    Specifications
    Material SMS (Spunbound Melt-Blown Synthetic)
    No. of Pockets 3
    Closure Type Snap
    Construction 3-ply SMS
    Size X-Large
    Disposable Yes
    Latex Free Yes
    Certifications/Compliance OSHA
    Style Knee length
    Thickness 3-ply
    Color Medium Blue
  • Description & Specifications
Related Products
Product Content Feedback