Learn More
ValueMax™ Extra-Safe™ Lab Coats — Purple
Static-free — no clinging
Supplier:  ValueMax™ 3660PPS
 View more versions of this product
Catalog No. 23-900-514A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Knee length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Small
3-ply SMS
Snaps
Knee length
Yes
OSHA
SMS
Purple
Yes
3
3-ply