Learn More
ValueMax™ Extra-Safe™ Lab Coats — Raspberry
Static-free — no clinging
Specifications
Certifications/Compliance OSHA
Closure Type Snaps
Color Raspberry
Construction 3-ply SMS
Material SMS
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-517E
View Documents
ValueMax™
3660RB2XL
2X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-517B
View Documents
ValueMax™
3660RBM
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-517H
View Documents
ValueMax™
3660RB5XL
5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-517F
View Documents
ValueMax™
3660RB3XL
3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-517D
View Documents
ValueMax™
3660RBXL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-517A
View Documents
ValueMax™
3660RBS
Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-517C
View Documents
ValueMax™
3660RBL/DEL
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Knee length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
OSHA
Raspberry
SMS
Yes
Knee length
Snaps
3-ply SMS
3
Yes
3-ply