ValuMax™ Extra-Safe™ Lab Coats — Sky Blue
Static-free — no clinging
Manufacturer: Valumax International, Inc 3660SB2XL
 View more versions of this product
Catalog No. 23-900-516E
  Check Availability
Add to Cart
    Specifications
    Material SMS
    No. of Pockets 3.0
    Size 2X-Large
    Disposable Yes
    Latex Free Yes
    Certifications/Compliance OSHA
    Style Knee length
    Thickness 3-ply
    Closure Type Snap Front
    Color Sky Blue
    Construction 3-ply SMS
  • Description & Specifications
Related Products
Product Content Feedback