ValuMax™ Extra-Safe™ Lab Coats — Teal
Static-free — no clinging
Manufacturer: Valumax International, Inc 3660TE5XL
 View more versions of this product
Catalog No. 23-900-511H
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance OSHA
    Closure Type Snap Front
    Color Teal
    Construction 3-ply SMS
    Material SMS
    No. of Pockets 3.0
    Size 5X-Large
    Disposable Yes
    Latex Free Yes
    Style Knee length
    Thickness 3-ply
    Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
    • Fluid-resistant, breathable
    • Snap front, three pockets, knit cuffs and collar
    • Knee length
    • Autoclavable and washable up to five times
    • Latex-free, fire retardant
  • Description & Specifications
Related Products
Product Content Feedback