Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Blueberry
Click to view available options
Large
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid-resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Hip length
- Autoclavable and washable up to five times
- Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Size | Large |
Material | Polypropylene |
Color | Blue |
No. of Pockets | 3 |
Latex Free | Yes |