Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Blueberry
Static-free – no clinging
Specifications
Color Blue
Material Polypropylene
No. of Pockets 3
Latex Free Yes
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-510G
View Documents
ValueMax™
3630BB3XL
3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-510F
View Documents
ValueMax™
3630BB2XL
2X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-510D
View Documents
ValueMax™
3630BBL
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-510I
View Documents
ValueMax™
3630BB5XL
5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-510B
View Documents
ValueMax™
3630BBS
Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-510C
View Documents
ValueMax™
3630BBM
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-510E
View Documents
ValueMax™
3630BBXL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Blue
Polypropylene
3
Yes