Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Cranberry
Static free — no clinging
Specifications
Color Cranberry
No. of Pockets 3
Latex Free Yes
View More Specs
Products 9
Catalog Number Mfr. No. Material Size Price Quantity & Availability  
Catalog Number Mfr. No. Material Size Price Quantity & Availability  
23-900-501I
View Documents
ValueMax™
3630CY5XL
Polypropylene 5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501G
View Documents
ValueMax™
3630CY3XL
Polypropylene 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501F
View Documents
ValueMax™
3630CY2XL
Polypropylene 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501D
View Documents
ValueMax™
3630CYL
Polypropylene Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501E
View Documents
ValueMax™
3630CYXL
Polypropylene X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501B
View Documents
ValueMax™
3630CYS
SMS Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501C
View Documents
ValueMax™
3630CYM
Polypropylene Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501H
View Documents
ValueMax™
3630CY4XL
Polypropylene 4X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-501A
View Documents
ValueMax™
3630CYXS
SMS X-Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Cranberry
3
Yes