Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Medical Blue
Static-free – no clinging
Supplier: ValueMax™ 3630MB3XL
This item is not returnable.
View return policy
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid-resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Hip length
- Autoclavable and washable up to five times
- Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
3X-Large | |
Blue | |
Yes |
Polypropylene | |
3 |