Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Medical Blue
Static-free – no clinging
Specifications
Material Polypropylene
Color Blue
No. of Pockets 3
Latex Free Yes
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-509I
View Documents
ValueMax™
3630MB5XL
5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
23-900-509G
View Documents
ValueMax™
3630MB3XL
3X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
23-900-509A
View Documents
ValueMax™
3630MBXS
X-Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Polypropylene
Blue
3
Yes