Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Multicolored
Static free — no clinging
Specifications
Color Multi-colored
Material Polypropylene
No. of Pockets 3
Latex Free Yes
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-504H
View Documents
ValueMax™
3630PGO4XL
4X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504D
View Documents
ValueMax™
3630PGOL
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504I
View Documents
ValueMax™
3630PGO5XL
5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
23-900-504F
View Documents
ValueMax™
3630PGO2XL
2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504G
View Documents
ValueMax™
3630PGO3XL
3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504C
View Documents
ValueMax™
3630PGOM
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504B
View Documents
ValueMax™
3630PGOS
Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504E
View Documents
ValueMax™
3630PGOXL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-504A
View Documents
ValueMax™
3630PGOXS
X-Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Multi-colored
Polypropylene
3
Yes