Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Purple
Static free — no clinging
$27.64
Specifications
Color Purple
Material Polypropylene
No. of Pockets 3
Latex Free Yes
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-503G
View Documents
ValueMax™
3630PP3XL
3X-Large
Pack of 10 for $27.64
Case of 10 PK for $276.41
Add to cart
23-900-503F
View Documents
ValueMax™
3630PP2XL
2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503D
View Documents
ValueMax™
3630PPL
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503H
View Documents
ValueMax™
3630PP4XL
4X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503I
View Documents
ValueMax™
3630PP5XL
5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503C
View Documents
ValueMax™
3630PPM
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503B
View Documents
ValueMax™
3630PPS
Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503A
View Documents
ValueMax™
3630PPXS
X-Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-503E
View Documents
ValueMax™
3630PPXL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Purple
Polypropylene
3
Yes