Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Raspberry
Static-free — no clinging
Manufacturer:  ValueMax™ 3630RB2XL
Catalog No. 23-900-506F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
2X-Large
Raspberry
Yes
Polypropylene
3