Learn More
ValueMax Extra-Safe Lab Jackets Raspberry
Static-free no clinging
Specifications
Color | Raspberry |
---|---|
Material | Polypropylene |
No. of Pockets | 3 |
Latex Free | Yes |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
23-900-506G
|
ValueMax
3630RB3XL |
3X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506F
|
ValueMax
3630RB2XL |
2X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506C
|
ValueMax
3630RBM |
Medium |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506I
|
ValueMax
3630RB5XL |
5X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506D
|
ValueMax
3630RBL |
Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506B
|
ValueMax
3630RBS |
Small |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506E
|
ValueMax
3630RBXL |
X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-900-506A
|
ValueMax
3630RBXS |
X-Small |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
- Fluid-resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Hip length
- Autoclavable and washable up to five times
- Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Raspberry | |
Polypropylene | |
3 | |
Yes |