Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
ValueMax™ Extra-Safe™ lab Jackets — Sky Blue
Static free — no clinging
$34.25
Specifications
Color Blue
Material Polypropylene
No. of Pockets 3
Latex Free Yes
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-505I
View Documents
ValueMax™
3630SB5XL
5X-Large
Pack of 10 for $34.25
Case of 10 PK for $342.50
N/A
23-900-505C
View Documents
ValueMax™
3630SBM
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505H
View Documents
ValueMax™
3630SB4XL
4X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505G
View Documents
ValueMax™
3630SB3XL
3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505F
View Documents
ValueMax™
3630SB2XL
2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505D
View Documents
ValueMax™
3630SBL
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505B
View Documents
ValueMax™
3630SBS
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505E
View Documents
ValueMax™
3630SBXL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-505A
View Documents
ValueMax™
3630SBXS
X-Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
Blue
Polypropylene
3
Yes