ValuMax™ Extra-Safe™ Lab Jackets — Teal
Static free — no clinging
Manufacturer: Valumax International, Inc 3630TEL
 View more versions of this product
Catalog No. 23-900-500D
  Check Availability
Add to Cart
    Specifications
    Material SMS
    No. of Pockets 3.0
    Size Large
    Disposable Yes
    Latex Free Yes
    Certifications/Compliance OSHA
    Style Hip length
    Thickness 3-ply
    Closure Type Snap Front
    Color Teal
    Construction 3-ply SMS
  • Description & Specifications
Related Products
Product Content Feedback