Learn More
ValueMax™ Extra-Safe™ Lab Jackets — White
Static-free — no clinging
Specifications
Color White
Material Polypropylene
No. of Pockets 3
Latex Free Yes
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
23-900-508G
View Documents
ValueMax™
3630WH3XL
3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508F
View Documents
ValueMax™
3630WH2XL
2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508H
View Documents
ValueMax™
3630WH4XL
4X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508C
View Documents
ValueMax™
3630WHM
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508I
View Documents
ValueMax™
3630WH5XL
5X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508D
View Documents
ValueMax™
3630WHL
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
23-900-508B
View Documents
ValueMax™
3630WHS
Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508E
View Documents
ValueMax™
3630WHXL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
23-900-508A
View Documents
ValueMax™
3630WHXS
X-Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Specifications
White
Polypropylene
3
Yes