Learn More
Valutek™ Cleanroom Paper
Specially formulated to minimize particulation in controlled environments
Manufacturer:  VALUE TEK VTCRPS23G8511
 View more versions of this product
Catalog No. 19-150-1187
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Synthetic Polymer
11 x 8.5 in. (279 x 216mm)
Laser printers and copy machines
279mm
10.4kg
216mm
Cleanroom Paper
Green
To minimize particulation in controlled environments
11 in.
23 lb.
8.5 in.
ISO 4 (class 10)