Learn More
Chemglass Life Sciences Valve, 0-4mm, S-Type, Exposed O-Ring Tip
SDP
Supplier:  Chemglass Life Sciences CGK52701
Encompass_Preferred
Valve, 0-4mm, S-Type, Exposed O-Ring Tip. Hi-vac valve with two sidearms sealed to the barrel at a 180° angle to one another and on the same plane.
Catalog No. 50-200-706