Learn More
Leap PAL PartsSupplier Diversity Partner VICI Valco Valve, 6 Port, Valco, 5000 PSI, 1.7mm Bore, HPLC Preparative (VAL.DL6UWK-CTC)  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner PAL.DL6UW
VICI Valco Valve, 6 Port, VICI Valco, 5000 PSI, 1.7mm Bore, HPLC Preparative (NEW LPP PN: VAL.DL6UWK-CTC). Made in the USA.
Catalog No. 50-635-014