Learn More
Brady™ Valve Lockout Pouch Kit with Brady Steel Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Valve Lockout Pouch Kit is an economical valve lockout pouch kit is perfect for locking out small gate and ball valves. A successful program begins with the right devices in the right place.
Manufacturer:  Brady™ 99681
Catalog No. 19-104-201
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Mechanical risk lockouts
8 in.
3 in.
7 in.
Safety
White on red
(1) lockout bag; (1) adjustable gate valve lockout, 1-6.5 in.; (2) small ball valve lockouts; (1) base cover plug valve lockout, 7/8 in.; (1) base cover plug valve lockout, 1-3/8 in.; (2) kd steel padlocks, 0.75 in. shackle, red; (2) lockout tags; (2) nylon cable ties
Nylon
3 in. x 7 in. x 8 in.