Learn More
Brady™ Valve Lockout Toolbox Kit w/ Brady Steel Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Valve Lockout Toolbox Kit is a convienent kit that provides almost everything needed to lock out gate, ball, butterfly and other valves.
Manufacturer:  Brady™ 99324
Catalog No. 19-104-195
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Mechanical risk lockouts
7.5 in.
8 in.
14 in.
Safety
Gray
(1) lockout box; (1) small universal lockout w/ arm; (1) small ball valve lockout; (1) gate valve lockout, 1-2.5 in.; (1) adjustable gate valve lockout, 1-6.5 in.; (2) kd steel padlocks, 0.75 in. shackle, red; (2) lockout tags; (1) all purpose cable lockout with 8 ft sheathed cable
Polyethylene
8 in. x 14 in. x 7.5 in.