Learn More
Leap PAL PartsSupplier Diversity Partner VICI Valco 10 Port Cheminert Injection Valve, 0.40mm, PEEK (VAL.C2V-2340D-CTC)  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner PAL.VLV10PK
VICI Valco Valve 10 Port 2 Position, 5K PSI, 0.40mm Ports, PEEK, Cheminert [VICI Valco PN: C2V-2340D-CTC] (NEW LPP PN: VAL.C2V-2340D-CTC). Thermo Part Number: 00950-01-00331. Made in the USA.
Catalog No. 50-635-033