Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAM1 Mouse anti-Human, DyLight™ 680, Clone: OTI3H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274824FR

Catalog No. NBP274824FR


Add to cart

Description

Description

VAM1 Monoclonal antibody specifically detects VAM1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

VAM1
OTI3H5
Western Blot
Affinity Purified, Conjugated
MPP6
Full length recombinant protein of human VAM1 (NP_057531) produced in E.coli.
Immunogen affinity purified
Stem Cell Markers
Primary
51678.0
Western Blot
DyLight 680
PBS with 0.05% Sodium Azide
MAGUK protein p55T, membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6), p55T, protein associated with Lin7 2, VAM-1PALS2VAM1MAGUK p55 subfamily member 6, Veli-associated MAGUK 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit