Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP-5 Mouse anti-Human, Janelia Fluor 646, Clone: 300000, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP250357JF646
Description
VAMP-5 Monoclonal antibody specifically detects VAMP-5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
VAMP-5 | |
300000 | |
Immunocytochemistry/Immunofluorescence | |
Purified | |
VAMP5 | |
GST-mV5-II and KLH-mV5-II peptide sequences | |
Protein G purified | |
Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission | |
Primary | |
10791 |
Immunocytochemistry, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin) | |
Mouse | |
IgG2a | |
0.1mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title