Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VAMP5 Monoclonal Antibody (3D4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524435
Description
Peptide Sequence: MAGIELERCQ QQANEVTEIM RNNFGKVLER GVKLAELQQR SDQLLDMSST FNKTTQNLAQ KKCWENIRYR.
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis.
Specifications
VAMP5 | |
Monoclonal | |
Unconjugated | |
Vamp5 | |
AF119384; Camp; HSPC191; Myobrevin; vamp; Vamp5; VAMP-5; vesicle associated membrane protein 5; vesicle-associated membrane protein 5 | |
Mouse | |
Protein A | |
RUO | |
10791 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3D4 | |
PBS with no preservative; pH 7.4 | |
O95183 | |
Vamp5 | |
VAMP5 (NP_006625, 1 a.a. ~ 70 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction