Learn More
Aviva Systems Biology Corp VAMP7 Antibody - N-terminal region (ARP47594_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47594_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VAMP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VAMP7. Alternate names: SYBL1, TI-VAMP, VAMP-7, TIVAMPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59601