Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA589528
Description
Immunogen sequence: MEEASEGGGN DRVRNLQSEV EGVKNIMTQN VERILARGEN LEHLRNKTED LEATSEHFKT TSQKVARKFW WKNVK; Positive Samples: HeLa, NIH/3T3, PC-12, Mouse kidney, Mouse liver, Rat lung; Cellular Location: Cell membrane, Early endosome membrane, Late endosome membrane, Lysosome membrane, Single-pass type IV membrane protein
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.Specifications
VAMP8 | |
Polyclonal | |
Unconjugated | |
VAMP8 | |
Vamp8 | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-75 of human VAMP8 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
2.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70404, Q9BV40, Q9WUF4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22320, 83730, 8673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title