Learn More
VAMP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155461AP
Catalog No. 50-173-0304
Add to cart
Description
Description
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
Specifications
Specifications
VAMP8
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70404, Q9BV40, Q9WUF4
EDB, Endobrevin, VAMP 8, VAMP8
Rabbit
IgG
150 μL
-20°C
Primary
22320, 83730, 8673
Western Blot
Unconjugated
VAMP8
Liquid
VAMP8
VAMP8 Fusion Protein Ag7903
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat