Learn More
Van de Graaff Generator, Jr.
SCI_ED
Includes: Instructions, 10cm Aluminum sphere mounted on top of a clear Plastic column, 4 in. dia. electrode
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S43200
SCI_ED
View Documents
Science First
N105S
Electrostatic Generator This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S43201
SCI_ED
View Documents
Science First
RP620
Transport Belt This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Use this junior model of the Van de Graaff generator for numerous electrostatic experiments and displays.
This 34cm mini version produces 75,000 volts and delivers sparks up to 10cm in length. The metal base houses an electric motor that operates on 115VAC 50/60Hz. The 10cm aluminum sphere sits on top of a clear plastic column that allows students to see the moving belt that delivers charge to the sphere.
Order Info
Replacement belts measuring 2W x 28cmL are also available separately.