Alfa Aesar™ Van Dyke Brown

Manufacturer: Alfa Aesar™ A1229314

Quantity 25g

 View more versions of this product

Catalog No. AAA1229314

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 72669-22-8
    MDL Number MFCD00148765
    Synonym Natural Brown 8
    Chemical Name or Material Van Dyke Brown

    Specifications

    Quantity 25g
    CAS 72669-22-8
    MDL Number MFCD00148765
    Synonym Natural Brown 8
    Chemical Name or Material Van Dyke Brown

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit