Learn More
Vanadium carbide, 99% (metals basis), Thermo Scientific Chemicals
Description
Vanadium carbide is used as an additive to tungsten carbide to fine the carbide crystals to improve the property of the cermets. It used in cutting tools and the steel industry. Used as material for research in electronics. Used anode materials for lithium ion batteries, and in supercapacitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2810°C |
Color | Black |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99% metal basis |
Solubility Information | Insoluble in water. |
Formula Weight | 62.95 |
Odor | Odorless |
Chemical Name or Material | Vanadium carbide |
Safety and Handling
Hazard Category | H228 |
Precautionary Statement | P210-P240-P241-P280-P370+P378q |
DOTInformation | Hazard Class: 4.1; Packaging Group: III |
EINECSNumber | 235-122-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.